Product Details

SNP ID
rs145600139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:64225042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTTGTAGTCAGGGTCTGCCCC[A/G]TGGAGCAAAAGCAGGCGGAAACAGT
Phenotype
MIM: 300891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB12 PubMed Links

Gene Details

Gene
ASB12
Gene Name
ankyrin repeat and SOCS box containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130388.3 802 Silent Mutation CAC,CAT H203H NP_569059.3

View Full Product Details