Product Details

SNP ID
rs145715341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:84064396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGGCAGAGTATGTTGCAACCA[C/T]TGCTCCCTAAAGTAATGAGAAAATG
Phenotype
MIM: 300303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS6KA6 PubMed Links

Gene Details

Gene
RPS6KA6
Gene Name
ribosomal protein S6 kinase A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014496.4 2104 Missense Mutation ATG,GTG M707V NP_055311.1
XM_011530917.2 2104 Missense Mutation ATG,GTG M707V XP_011529219.1
XM_011530918.2 2104 Missense Mutation ATG,GTG M707V XP_011529220.1
XM_011530919.2 2104 Missense Mutation ATG,GTG M604V XP_011529221.1
XM_011530920.2 2104 Missense Mutation ATG,GTG M586V XP_011529222.1
XM_017029423.1 2104 Missense Mutation ATG,GTG M724V XP_016884912.1
XM_017029424.1 2104 Missense Mutation ATG,GTG M724V XP_016884913.1
XM_017029425.1 2104 Missense Mutation ATG,GTG M724V XP_016884914.1
XM_017029426.1 2104 Missense Mutation ATG,GTG M468V XP_016884915.1

View Full Product Details