Product Details

SNP ID
rs145759402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:19929605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAAAGCATTTTAAGGTTACACAG[C/T]CTCACAACTGTATTACCATTACTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf23 PubMed Links

Gene Details

Gene
CXorf23
Gene Name
chromosome X open reading frame 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198279.3 Intron NP_938020.2
XM_005274473.3 Intron XP_005274530.1
XM_005274474.3 Intron XP_005274531.1
XM_005274475.3 Intron XP_005274532.1
XM_011545475.2 Intron XP_011543777.1
XM_011545476.2 Intron XP_011543778.1
XM_011545477.2 Intron XP_011543779.1
XM_011545478.2 Intron XP_011543780.1
XM_011545479.2 Intron XP_011543781.1
XM_011545482.2 Intron XP_011543784.1
XM_017029390.1 Intron XP_016884879.1
XM_017029391.1 Intron XP_016884880.1
XM_017029392.1 Intron XP_016884881.1

View Full Product Details