Product Details

SNP ID
rs145815177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:63638070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACCTGCTGAAGTTTTGCCAGAA[A/T]GGTGACTGGCTGCGCTTGGGTTCGG
Phenotype
MIM: 300429
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF9 PubMed Links

Gene Details

Gene
ARHGEF9
Gene Name
Cdc42 guanine nucleotide exchange factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173479.1 2437 Silent Mutation CCA,CCT P450P NP_001166950.1
NM_001173480.1 2437 Silent Mutation CCA,CCT P401P NP_001166951.1
NM_015185.2 2437 Silent Mutation CCA,CCT P503P NP_056000.1
XM_005262249.2 2437 Silent Mutation CCA,CCT P510P XP_005262306.1
XM_005262250.3 2437 Silent Mutation CCA,CCT P482P XP_005262307.1
XM_005262251.1 2437 Silent Mutation CCA,CCT P482P XP_005262308.1
XM_005262252.1 2437 Silent Mutation CCA,CCT P482P XP_005262309.1
XM_011530890.1 2437 Silent Mutation CCA,CCT P503P XP_011529192.1
XM_011530892.1 2437 Silent Mutation CCA,CCT P443P XP_011529194.1
XM_017029363.1 2437 Silent Mutation CCA,CCT P508P XP_016884852.1
XM_017029364.1 2437 Silent Mutation CCA,CCT P508P XP_016884853.1
XM_017029365.1 2437 Silent Mutation CCA,CCT P503P XP_016884854.1
XM_017029366.1 2437 Silent Mutation CCA,CCT P503P XP_016884855.1
XM_017029367.1 2437 Silent Mutation CCA,CCT P482P XP_016884856.1
XM_017029368.1 2437 Silent Mutation CCA,CCT P482P XP_016884857.1
XM_017029369.1 2437 Silent Mutation CCA,CCT P466P XP_016884858.1
XM_017029370.1 2437 Silent Mutation CCA,CCT P459P XP_016884859.1
XM_017029371.1 2437 Silent Mutation CCA,CCT P438P XP_016884860.1
XM_017029372.1 2437 Silent Mutation CCA,CCT P438P XP_016884861.1
XM_017029373.1 2437 Silent Mutation CCA,CCT P401P XP_016884862.1
XM_017029374.1 2437 Silent Mutation CCA,CCT P399P XP_016884863.1
XM_017029375.1 2437 UTR 3 XP_016884864.1
XM_017029376.1 2437 Intron XP_016884865.1
XM_017029377.1 2437 UTR 3 XP_016884866.1
XM_017029378.1 2437 Intron XP_016884867.1
XM_017029379.1 2437 UTR 3 XP_016884868.1
XM_017029380.1 2437 UTR 3 XP_016884869.1

View Full Product Details