Product Details

SNP ID
rs145837656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49173411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTTCTTCCGAACCCTCATAGC[A/G]GCAATCCTCTACCTGATCACCTCCA
Phenotype
MIM: 300112 MIM: 300111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGIX PubMed Links

Gene Details

Gene
MAGIX
Gene Name
MAGI family member, X-linked
There are no transcripts associated with this gene.

Gene
PLP2
Gene Name
proteolipid protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002668.2 437 Silent Mutation GCA,GCG A91A NP_002659.1
Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
There are no transcripts associated with this gene.

View Full Product Details