Product Details

SNP ID
rs145902255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70043118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAGACCGGTTCTTCAACACCA[A/G]GGTAGAAGAACCTGGCAGGCTGTAT
Phenotype
MIM: 300925 MIM: 300451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AWAT2 PubMed Links

Gene Details

Gene
AWAT2
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002254.1 604 Silent Mutation CTG,TTG L200L NP_001002254.1
XM_011530876.2 604 Silent Mutation CTG,TTG L200L XP_011529178.1
Gene
EDA
Gene Name
ectodysplasin A
There are no transcripts associated with this gene.

View Full Product Details