Product Details

SNP ID
rs145922117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70448431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGTCTGCTGTCCTTCCAGTTC[A/G]GACACAGTGGGAGTCTATGATCCCC
Phenotype
MIM: 300189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLG3 PubMed Links

Gene Details

Gene
DLG3
Gene Name
discs large MAGUK scaffold protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166278.1 Intron NP_001159750.1
NM_020730.2 Intron NP_065781.1
NM_021120.3 Intron NP_066943.2
XM_005262248.3 Intron XP_005262305.1
XM_006724625.2 Intron XP_006724688.1
XM_006724626.2 Intron XP_006724689.1
XM_011530883.1 Intron XP_011529185.1
XM_017029322.1 Intron XP_016884811.1
XM_017029323.1 Intron XP_016884812.1
XM_017029324.1 Intron XP_016884813.1
XM_017029325.1 Intron XP_016884814.1
XM_017029326.1 Intron XP_016884815.1
XM_017029327.1 Intron XP_016884816.1
Gene
DLG3-AS1
Gene Name
DLG3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details