Product Details

SNP ID
rs145931176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:52648261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCCCGAATTCTTTCCTCTTAC[C/G]GGATGTCTTGTTAATCTTCTCAGAG
Phenotype
MIM: 300542
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SSX7 PubMed Links

Gene Details

Gene
SSX7
Gene Name
SSX family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173358.2 Intron NP_775494.1
XM_017029430.1 Intron XP_016884919.1
XM_017029431.1 Intron XP_016884920.1

View Full Product Details