Product Details

SNP ID
rs145969952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107067605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTTGGAGATTTGACCGTTGCTT[C/T]TTGTTCTTTCCAAACTCTATCACCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM41 PubMed Links

Gene Details

Gene
RBM41
Gene Name
RNA binding motif protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171080.1 1290 Silent Mutation AAA,AAG K388K NP_001164551.1
NM_001324242.1 1290 Silent Mutation AAA,AAG K412K NP_001311171.1
NM_001324243.1 1290 Silent Mutation AAA,AAG K447K NP_001311172.1
NM_001324244.1 1290 Silent Mutation AAA,AAG K412K NP_001311173.1
NM_018301.4 1290 Silent Mutation AAA,AAG K388K NP_060771.3
XM_011530980.2 1290 Silent Mutation AAA,AAG K447K XP_011529282.1
XM_011530981.2 1290 Silent Mutation AAA,AAG K423K XP_011529283.1

View Full Product Details