Product Details
- SNP ID
-
hCV175487788
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:154380364 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCAGATGCTGACGCTTTCCATCA[C/T]CAGGTGAGCTGGCTGGCAGGCGTCC
- Phenotype
-
MIM: 300384
MIM: 300017
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EMD
PubMed Links
Gene Details
- Gene
- EMD
- Gene Name
- emerin
There are no transcripts associated with this gene.
- Gene
- FLNA
- Gene Name
- filamin A
There are no transcripts associated with this gene.
- Gene
- LOC107987332
- Gene Name
- uncharacterized LOC107987332
There are no transcripts associated with this gene.
View Full Product Details