Product Details

SNP ID
rs146310467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77959979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCCTGTTTCTCCATATCCTCAC[C/T]GACACTTGGTATTGTCAGATTTTGA
Phenotype
MIM: 300011 MIM: 300567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP7A PubMed Links

Gene Details

Gene
ATP7A
Gene Name
ATPase copper transporting alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000052.6 Intron NP_000043.4
NM_001282224.1 Intron NP_001269153.1
Gene
PGAM4
Gene Name
phosphoglycerate mutase family member 4
There are no transcripts associated with this gene.

View Full Product Details