Product Details
- SNP ID
-
rs146330434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:18994981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTACTGCTTGACTGTAAGGAATT[C/T]GAAGAGCTGGACACTCCTTGGTTTA
- Phenotype
-
MIM: 300572
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRG2
PubMed Links
Gene Details
- Gene
- ADGRG2
- Gene Name
- adhesion G protein-coupled receptor G2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001079858.2 |
2912 |
Silent Mutation |
TCA,TCG |
S928S |
NP_001073327.1 |
NM_001079859.2 |
2912 |
Silent Mutation |
TCA,TCG |
S914S |
NP_001073328.1 |
NM_001079860.2 |
2912 |
Silent Mutation |
TCA,TCG |
S906S |
NP_001073329.1 |
NM_001184833.1 |
2912 |
Silent Mutation |
TCA,TCG |
S912S |
NP_001171762.1 |
NM_001184834.1 |
2912 |
Intron |
|
|
NP_001171763.1 |
NM_001184835.1 |
2912 |
Silent Mutation |
TCA,TCG |
S890S |
NP_001171764.1 |
NM_001184836.1 |
2912 |
Silent Mutation |
TCA,TCG |
S904S |
NP_001171765.1 |
NM_001184837.1 |
2912 |
Silent Mutation |
TCA,TCG |
S898S |
NP_001171766.1 |
NM_005756.3 |
2912 |
Silent Mutation |
TCA,TCG |
S925S |
NP_005747.2 |
XM_006724455.3 |
2912 |
Silent Mutation |
TCA,TCG |
S928S |
XP_006724518.1 |
XM_011545434.1 |
2912 |
Silent Mutation |
TCA,TCG |
S928S |
XP_011543736.1 |
XM_011545435.2 |
2912 |
Silent Mutation |
TCA,TCG |
S928S |
XP_011543737.1 |
- Gene
- LOC101928415
- Gene Name
- uncharacterized LOC101928415
There are no transcripts associated with this gene.
View Full Product Details