Product Details

SNP ID
rs146468682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24695026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTATCAGTGATTTGCCACTAAG[A/C]CCTTTCTTCATCCTAAAAACTACGT
Phenotype
MIM: 312040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POLA1 PubMed Links

Gene Details

Gene
POLA1
Gene Name
DNA polymerase alpha 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016937.3 Intron NP_058633.2
XM_005274552.3 Intron XP_005274609.1
XM_006724499.2 Intron XP_006724562.1
XM_011545540.2 Intron XP_011543842.1
XM_011545541.2 Intron XP_011543843.1
XM_017029594.1 Intron XP_016885083.1
XM_017029595.1 Intron XP_016885084.1
XM_017029596.1 Intron XP_016885085.1

View Full Product Details