Product Details

SNP ID
rs146484786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134243948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAAAGCTGTACATTAAGGATGT[A/G]CAGCTTTGGGTAGGGAAGACTCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC160 PubMed Links

Gene Details

Gene
CCDC160
Gene Name
coiled-coil domain containing 160
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101357.1 Intron NP_001094827.1
XM_005262409.4 Intron XP_005262466.1
XM_005262410.4 Intron XP_005262467.1

View Full Product Details