Product Details
- SNP ID
-
rs146572363
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:53194180 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACTGCCCTCTCTGAGGGGGTCAG[A/C]AGCAGGGGCTGCAGGGTAGTTAGGA
- Phenotype
-
MIM: 314690
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KDM5C
PubMed Links
Gene Details
- Gene
- KDM5C
- Gene Name
- lysine demethylase 5C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146702.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1266S |
NP_001140174.1 |
NM_001282622.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1332S |
NP_001269551.1 |
NM_004187.3 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
NP_004178.2 |
XM_005262035.4 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
XP_005262092.1 |
XM_011530824.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1307S |
XP_011529126.1 |
XM_011530825.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1292S |
XP_011529127.1 |
XM_011530826.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1292S |
XP_011529128.1 |
XM_011530827.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
XP_011529129.1 |
XM_011530828.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
XP_011529130.1 |
XM_011530829.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1168S |
XP_011529131.1 |
XM_011530830.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1168S |
XP_011529132.1 |
XM_011530831.2 |
4265 |
Missense Mutation |
GCT,TCT |
A1005S |
XP_011529133.1 |
XM_017029886.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1332S |
XP_016885375.1 |
XM_017029887.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
XP_016885376.1 |
XM_017029888.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1332S |
XP_016885377.1 |
XM_017029889.1 |
4265 |
Missense Mutation |
GCT,TCT |
A1333S |
XP_016885378.1 |
- Gene
- MIR6894
- Gene Name
- microRNA 6894
There are no transcripts associated with this gene.
- Gene
- MIR6895
- Gene Name
- microRNA 6895
There are no transcripts associated with this gene.
View Full Product Details