Product Details

SNP ID
rs146662539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30304758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCGTCATCCTGGTGTGTTCAC[C/T]GAGTATTTGCTGAGTTCCCCACTGG
Phenotype
MIM: 300473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NR0B1 PubMed Links

Gene Details

Gene
NR0B1
Gene Name
nuclear receptor subfamily 0 group B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000475.4 1249 Missense Mutation AGT,GGT S412G NP_000466.2
XM_017029338.1 1249 Intron XP_016884827.1

View Full Product Details