Product Details

SNP ID
rs146746942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107069302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTCATTCGTCCAGTCATCATT[C/T]GGAATTGAATTGGAGGTCCTTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM41 PubMed Links

Gene Details

Gene
RBM41
Gene Name
RNA binding motif protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171080.1 1154 Missense Mutation CAA,CGA Q343R NP_001164551.1
NM_001324242.1 1154 Missense Mutation CAA,CGA Q367R NP_001311171.1
NM_001324243.1 1154 Missense Mutation CAA,CGA Q402R NP_001311172.1
NM_001324244.1 1154 Missense Mutation CAA,CGA Q367R NP_001311173.1
NM_018301.4 1154 Missense Mutation CAA,CGA Q343R NP_060771.3
XM_011530980.2 1154 Missense Mutation CAA,CGA Q402R XP_011529282.1
XM_011530981.2 1154 Missense Mutation CAA,CGA Q378R XP_011529283.1

View Full Product Details