Product Details

SNP ID
rs146784489
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77456068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAGATTTCAACTAAAACATTT[C/T]TGTGTAACTCATGAAAGAGAATGAA
Phenotype
MIM: 309630 MIM: 300827
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FGF16 PubMed Links

Gene Details

Gene
FGF16
Gene Name
fibroblast growth factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003868.2 Intron NP_003859.1

View Full Product Details