Product Details

SNP ID
hCV175513827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153454463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCTGCTTCTTCACGGTCTCCA[C/T]GGCCTTCGCAGAGGTGTCAGCAACT
Phenotype
MIM: 300540 MIM: 300370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAUS7 PubMed Links

Gene Details

Gene
HAUS7
Gene Name
HAUS augmin like complex subunit 7
There are no transcripts associated with this gene.

Gene
TREX2
Gene Name
three prime repair exonuclease 2
There are no transcripts associated with this gene.

View Full Product Details