Product Details

SNP ID
rs147000148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:96690639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATTTGTCCCTTTGGAGGAATA[G/T]GCATTCTATCCTTTTACTAAGATTT
Phenotype
MIM: 300108
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DIAPH2 PubMed Links

Gene Details

Gene
DIAPH2
Gene Name
diaphanous related formin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006729.4 Intron NP_006720.1
NM_007309.3 Intron NP_009293.1

View Full Product Details