Product Details

SNP ID
rs147459801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71101132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGGGTGTCCCTCCCACCCTCA[A/G]TGAAGGTCTAGAGCTGTTAGATGGG
Phenotype
MIM: 300033 MIM: 308380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
There are no transcripts associated with this gene.

Gene
FOXO4
Gene Name
forkhead box O4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170931.1 1117 Missense Mutation AAT,AGT N246S NP_001164402.1
NM_005938.3 1117 Missense Mutation AAT,AGT N301S NP_005929.2
XM_011530957.2 1117 Missense Mutation AAT,AGT N123S XP_011529259.1
Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
There are no transcripts associated with this gene.

View Full Product Details