Product Details

SNP ID
rs147466639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:11118573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTAGAATTCACAATCAGAATAA[C/T]GAGCAGGCTTGGAAGGAGATTTTGA
Phenotype
MIM: 300056 MIM: 300552
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCCS PubMed Links

Gene Details

Gene
HCCS
Gene Name
holocytochrome c synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122608.2 691 Silent Mutation AAC,AAT N158N NP_001116080.1
NM_001171991.2 691 Silent Mutation AAC,AAT N158N NP_001165462.1
NM_005333.4 691 Silent Mutation AAC,AAT N158N NP_005324.3
Gene
MID1
Gene Name
midline 1
There are no transcripts associated with this gene.

View Full Product Details