Product Details

SNP ID
rs147569276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70426456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGCAGGAGTGGCATAGCTTGC[A/G]TGTGTCACTGCAGGTGAGTGGCCAA
Phenotype
MIM: 300521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GDPD2 PubMed Links

Gene Details

Gene
GDPD2
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171191.1 326 Missense Mutation CAT,CGT H71R NP_001164662.1
NM_001171192.1 326 Missense Mutation CAT,CGT H150R NP_001164663.1
NM_001171193.1 326 Missense Mutation CAT,CGT H71R NP_001164664.1
NM_017711.3 326 Missense Mutation CAT,CGT H150R NP_060181.2
XM_011530977.1 326 Missense Mutation CAT,CGT H23R XP_011529279.1
XM_017029614.1 326 UTR 5 XP_016885103.1
Gene
KIF4A
Gene Name
kinesin family member 4A
There are no transcripts associated with this gene.

Gene
LOC105373244
Gene Name
uncharacterized LOC105373244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531081.2 326 Intron XP_011529383.1
XM_011531082.2 326 Intron XP_011529384.1

View Full Product Details