Product Details

SNP ID
rs147637788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16124358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTTCACACTCACGGCGCTCTC[A/G]GCAGACAGGTAAGTACAGGCTGAAA
Phenotype
MIM: 305670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRPR PubMed Links

Gene Details

Gene
GRPR
Gene Name
gastrin releasing peptide receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005314.2 1058 Silent Mutation TCA,TCG S135S NP_005305.1

View Full Product Details