Product Details

SNP ID
rs147967317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88754129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTATTGTGAGGTCTGTGCTCTTT[A/G]TGTCACAGATACAAATTGAAAGTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 974 Missense Mutation ATG,GTG M239V NP_001171700.1
NM_033048.5 974 Missense Mutation ATG,GTG M239V NP_149037.4

View Full Product Details