Product Details

SNP ID
rs147976670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18996022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCTTTTCCTTGAGATTTAAGA[A/G]GAAGAGAAAAAGTGAGGTAAATCTT
Phenotype
MIM: 300572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRG2 PubMed Links

Gene Details

Gene
ADGRG2
Gene Name
adhesion G protein-coupled receptor G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079858.2 Intron NP_001073327.1
NM_001079859.2 Intron NP_001073328.1
NM_001079860.2 Intron NP_001073329.1
NM_001184833.1 Intron NP_001171762.1
NM_001184834.1 Intron NP_001171763.1
NM_001184835.1 Intron NP_001171764.1
NM_001184836.1 Intron NP_001171765.1
NM_001184837.1 Intron NP_001171766.1
NM_005756.3 Intron NP_005747.2
XM_006724455.3 Intron XP_006724518.1
XM_011545434.1 Intron XP_011543736.1
XM_011545435.2 Intron XP_011543737.1
Gene
LOC101928415
Gene Name
uncharacterized LOC101928415
There are no transcripts associated with this gene.

View Full Product Details