Product Details

SNP ID
hCV175549147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154402778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACAGTGAGGCTGAGAGGCTGGA[C/T]GCTGTGCGCCTGGCTCAGCTTCAGC
Phenotype
MIM: 300081 MIM: 312173 MIM: 300394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
There are no transcripts associated with this gene.

Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
There are no transcripts associated with this gene.

Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

Gene
TAZ
Gene Name
tafazzin
There are no transcripts associated with this gene.

View Full Product Details