Product Details

SNP ID
rs148432819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103077895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTTGGCTGGTACCCTCTAAAC[C/T]TTCCACAGAGCCCACCTTTGGAGAG
Phenotype
MIM: 300316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NXF3 PubMed Links

Gene Details

Gene
NXF3
Gene Name
nuclear RNA export factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022052.1 Intron NP_071335.1

View Full Product Details