Product Details

SNP ID
rs148444371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134543566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTTTTCCAAGTAACATTGACAC[A/G]CATCTGTTCAAATCAGTTGAGGTTC
Phenotype
MIM: 300682 MIM: 300865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00629 PubMed Links

Gene Details

Gene
LINC00629
Gene Name
long intergenic non-protein coding RNA 629
There are no transcripts associated with this gene.

Gene
MIR424
Gene Name
microRNA 424
There are no transcripts associated with this gene.

Gene
MIR450A1
Gene Name
microRNA 450a-1
There are no transcripts associated with this gene.

Gene
MIR450A2
Gene Name
microRNA 450a-2
There are no transcripts associated with this gene.

Gene
MIR450B
Gene Name
microRNA 450b
There are no transcripts associated with this gene.

Gene
MIR503
Gene Name
microRNA 503
There are no transcripts associated with this gene.

Gene
MIR503HG
Gene Name
MIR503 host gene
There are no transcripts associated with this gene.

Gene
MIR542
Gene Name
microRNA 542
There are no transcripts associated with this gene.

View Full Product Details