Product Details

SNP ID
rs148533678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55926596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATAATTTACTTTGCATATGGGGG[C/T]TAACACTTGGGATCAGGCCCTAGGG
Phenotype
MIM: 300286
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLF8 PubMed Links

Gene Details

Gene
KLF8
Gene Name
Kruppel like factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159296.2 Intron NP_001152768.1
NM_001324099.1 Intron NP_001311028.1
NM_001324100.1 Intron NP_001311029.1
NM_001324102.1 Intron NP_001311031.1
NM_001324104.1 Intron NP_001311033.1
NM_001324105.1 Intron NP_001311034.1
NM_007250.5 Intron NP_009181.2
XM_005261977.2 Intron XP_005262034.1
XM_005261979.3 Intron XP_005262036.1
XM_006724575.2 Intron XP_006724638.1
XM_006724576.3 Intron XP_006724639.1
XM_011530759.2 Intron XP_011529061.1
XM_017029250.1 Intron XP_016884739.1

View Full Product Details