Product Details

SNP ID
rs148562419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110014022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTGTTCCCACATTCATAAAA[C/T]GATATGATCAAACTTGAACATTACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM164 PubMed Links

Gene Details

Gene
TMEM164
Gene Name
transmembrane protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017698.2 3080 Intron NP_060168.2
NM_032227.3 3080 Intron NP_115603.2
XM_005262205.4 3080 Intron XP_005262262.1
XM_005262206.3 3080 Intron XP_005262263.1
XM_005262208.4 3080 Intron XP_005262265.1
XM_011531051.2 3080 Intron XP_011529353.1
XM_011531054.1 3080 Intron XP_011529356.1
XM_011531055.2 3080 Intron XP_011529357.1
XM_017029894.1 3080 Intron XP_016885383.1
XM_017029895.1 3080 UTR 3 XP_016885384.1
XM_017029896.1 3080 Intron XP_016885385.1
XM_017029897.1 3080 Intron XP_016885386.1
XM_017029898.1 3080 Intron XP_016885387.1
XM_017029899.1 3080 Intron XP_016885388.1
XM_017029900.1 3080 Intron XP_016885389.1

View Full Product Details