Product Details
- SNP ID
-
rs148609725
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:112779197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGCCGCTGCTGGAGCTGGAACC[A/G]GAACCAGAGCCGGAGCTGGAACTGG
- Phenotype
-
MIM: 300410
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMOT
PubMed Links
Gene Details
- Gene
- AMOT
- Gene Name
- angiomotin
- Gene
- MIR4329
- Gene Name
- microRNA 4329
There are no transcripts associated with this gene.
View Full Product Details