Product Details

SNP ID
rs148700490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55263593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATTGATGTATACAATGTGTTA[C/T]GAATAAGCAGGAGAAGGAAGCCATA
Phenotype
MIM: 300739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAGE3 PubMed Links

Gene Details

Gene
PAGE3
Gene Name
PAGE family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017931.2 Intron NP_001017931.2
NM_001171252.1 Intron NP_001164723.1
NM_001303613.1 Intron NP_001290542.1
XM_017029282.1 Intron XP_016884771.1

View Full Product Details