Product Details

SNP ID
rs148700620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120538205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTTGCTGGTGAAAGCAGCTCC[A/G]CATTCTATTAAAGATGTTTGTACAT
Phenotype
MIM: 300304
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL4B PubMed Links

Gene Details

Gene
CUL4B
Gene Name
cullin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079872.1 2181 Silent Mutation TGC,TGT C619C NP_001073341.1
NM_003588.3 2181 Silent Mutation TGC,TGT C637C NP_003579.3
XM_005262481.1 2181 Silent Mutation TGC,TGT C637C XP_005262538.1
XM_006724784.1 2181 Silent Mutation TGC,TGT C624C XP_006724847.1
XM_006724785.2 2181 Silent Mutation TGC,TGT C624C XP_006724848.1
XM_011531399.2 2181 Silent Mutation TGC,TGT C441C XP_011529701.1
XM_011531400.2 2181 Silent Mutation TGC,TGT C441C XP_011529702.1
XM_011531401.1 2181 Silent Mutation TGC,TGT C423C XP_011529703.1

View Full Product Details