Product Details

SNP ID
hCV175573348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135349185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATACCACCAACCATGCACCTA[C/G]AGTCACCTGCACTCCAGGTAATGGG
Phenotype
MIM: 300627 MIM: 314997
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF449 PubMed Links

Gene Details

Gene
ZNF449
Gene Name
zinc finger protein 449
There are no transcripts associated with this gene.

Gene
ZNF75D
Gene Name
zinc finger protein 75D
There are no transcripts associated with this gene.

View Full Product Details