Product Details

SNP ID
rs149118628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:126549569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACATTTACAACTGACTTGGAGA[C/T]GCATGCACACAAATATATCTTACTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF12L1 PubMed Links

Gene Details

Gene
DCAF12L1
Gene Name
DDB1 and CUL4 associated factor 12 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178470.4 3204 UTR 3 NP_848565.2

View Full Product Details