Product Details

SNP ID
rs149296012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50371763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGATTCATCCAGTCAATCTCAGA[A/T]AGCTTTTTGAACTCCTTATTCATGG
Phenotype
MIM: 300837
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 3658 Silent Mutation CTA,CTT L1191L NP_001013764.1
XM_017029268.1 3658 Silent Mutation CTA,CTT L1162L XP_016884757.1

View Full Product Details