Product Details

SNP ID
rs149384548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133026133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTTCACATATTTCTTTCGTTT[C/T]GGATTTTCGGGCACTGTTTGAAAGT
Phenotype
MIM: 300309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP26 PubMed Links

Gene Details

Gene
USP26
Gene Name
ubiquitin specific peptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031907.1 2226 Silent Mutation CCA,CCG P696P NP_114113.1
XM_017029892.1 2226 Silent Mutation CCA,CCG P696P XP_016885381.1

View Full Product Details