Product Details

SNP ID
rs149506702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154434327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATGACACCGCTCCCCGGATCCT[A/G]TTCTGGGCCCAAAACTTCTCTGTGG
Phenotype
MIM: 300197 MIM: 300453 MIM: 300104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6AP1 PubMed Links

Gene Details

Gene
ATP6AP1
Gene Name
ATPase H+ transporting accessory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001183.5 1195 Silent Mutation CTA,CTG L268L NP_001174.2
XM_011531179.1 1195 Silent Mutation CTA,CTG L79L XP_011529481.1
Gene
CH17-340M24.3
Gene Name
uncharacterized protein BC009467
There are no transcripts associated with this gene.

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
There are no transcripts associated with this gene.

Gene
GDI1
Gene Name
GDP dissociation inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details