Product Details

SNP ID
rs149817919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30559535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTTGACATCAATTGCAATAGG[C/T]GGCTAAAGACTATATCTCTGGCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 1216 Missense Mutation CAC,CGC H274R NP_079435.1

View Full Product Details