Product Details

SNP ID
rs149833963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2770588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTGGCAGATGCCCTTGATGAC[C/T]CTGGTAAGTGCCGATATTTCAAGGG
Phenotype
MIM: 300879
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
XG PubMed Links

Gene Details

Gene
XG
Gene Name
Xg blood group
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141919.1 1251 Intron NP_001135391.1
NM_001141920.1 1251 Intron NP_001135392.1
NM_175569.2 1251 Intron NP_780778.1
XM_005274587.3 1251 Missense Mutation CCT,TCT P34S XP_005274644.1
XM_011545575.2 1251 Missense Mutation CCT,TCT P34S XP_011543877.1
XM_017029787.1 1251 Intron XP_016885276.1

View Full Product Details