Product Details

SNP ID
rs149950630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153982246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCGTTCAACTCACTCGTGAAC[A/G]TGGCCTACACGCTGCTGGGGCTGTC
Phenotype
MIM: 300059
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3202-1 PubMed Links

Gene Details

Gene
MIR3202-1
Gene Name
microRNA 3202-1
There are no transcripts associated with this gene.

Gene
MIR3202-2
Gene Name
microRNA 3202-2
There are no transcripts associated with this gene.

Gene
TMEM187
Gene Name
transmembrane protein 187
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003492.2 733 Missense Mutation ATG,GTG M62V NP_003483.1
XM_011531199.2 733 Missense Mutation ATG,GTG M62V XP_011529501.1

View Full Product Details