Product Details

SNP ID
rs150013472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135905301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGAGAGGATGGAAAATGGCCA[A/G]TCTCGAACTGACAAAGTCTTGTCAA
Phenotype
MIM: 300359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAGE1 PubMed Links

Gene Details

Gene
SAGE1
Gene Name
sarcoma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018666.2 538 Silent Mutation CAA,CAG Q121Q NP_061136.2
XM_017029621.1 538 Silent Mutation CAA,CAG Q121Q XP_016885110.1
XM_017029622.1 538 Intron XP_016885111.1
XM_017029623.1 538 Silent Mutation CAA,CAG Q121Q XP_016885112.1
XM_017029624.1 538 Silent Mutation CAA,CAG Q90Q XP_016885113.1
XM_017029625.1 538 Silent Mutation CAA,CAG Q121Q XP_016885114.1
XM_017029626.1 538 Intron XP_016885115.1
XM_017029627.1 538 Intron XP_016885116.1

View Full Product Details