Product Details

SNP ID
rs150076182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:77550185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAATTCCAACACTGCTTTTCAAT[A/G]ATATATGGATCACTAAGGAACACTA
Phenotype
MIM: 300504 MIM: 300032
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRX PubMed Links

Gene Details

Gene
ATRX
Gene Name
ATRX, chromatin remodeler
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000489.4 Intron NP_000480.3
NM_138270.3 Intron NP_612114.2
XM_005262153.4 Intron XP_005262210.2
XM_005262154.4 Intron XP_005262211.2
XM_005262155.4 Intron XP_005262212.2
XM_005262156.4 Intron XP_005262213.2
XM_005262157.4 Intron XP_005262214.2
XM_006724666.3 Intron XP_006724729.1
XM_006724667.3 Intron XP_006724730.1
XM_006724668.2 Intron XP_006724731.1
XM_017029601.1 Intron XP_016885090.1
XM_017029602.1 Intron XP_016885091.1
XM_017029603.1 Intron XP_016885092.1
XM_017029604.1 Intron XP_016885093.1
XM_017029605.1 Intron XP_016885094.1
XM_017029606.1 Intron XP_016885095.1
XM_017029607.1 Intron XP_016885096.1
XM_017029608.1 Intron XP_016885097.1
XM_017029609.1 Intron XP_016885098.1
XM_017029610.1 Intron XP_016885099.1
XM_017029611.1 Intron XP_016885100.1

View Full Product Details