Product Details

SNP ID
rs150201638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53077006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTACATGGCCATCGCCCACCAC[C/T]GCTTCTACGCCAAGCGCATGACACT
Phenotype
MIM: 300253 MIM: 300564
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR173 PubMed Links

Gene Details

Gene
GPR173
Gene Name
G protein-coupled receptor 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018969.5 660 Missense Mutation CGC,TGC R129C NP_061842.1
XM_011530798.2 660 Missense Mutation CGC,TGC R129C XP_011529100.1
Gene
TSPYL2
Gene Name
TSPY like 2
There are no transcripts associated with this gene.

View Full Product Details