Product Details

SNP ID
rs150227672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153944131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGTCATCATACACCCGCCCCTC[A/G]CGGCCAAGGCACGTGAAGAAGCCCC
Phenotype
MIM: 300019 MIM: 300013 MIM: 312420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCFC1 PubMed Links

Gene Details

Gene
HCFC1
Gene Name
host cell factor C1
There are no transcripts associated with this gene.

Gene
NAA10
Gene Name
N(alpha)-acetyltransferase 10, NatA catalytic subunit
There are no transcripts associated with this gene.

Gene
RENBP
Gene Name
renin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002910.5 332 Silent Mutation CGC,CGT R54R NP_002901.2
XM_017029698.1 332 Silent Mutation CGC,CGT R44R XP_016885187.1

View Full Product Details