Product Details
- SNP ID
-
rs150246034
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135521064 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCGCACTGACCTGGGCACCTCTTA[C/T]TTGGACATTGCCAAAGGCGCTGTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
INTS6L
PubMed Links
Gene Details
- Gene
- INTS6L
- Gene Name
- integrator complex subunit 6 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182540.4 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
NP_872346.3 |
XM_005262394.2 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_005262451.1 |
XM_006724740.2 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_006724803.1 |
XM_006724741.3 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_006724804.1 |
XM_011531311.2 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_011529613.1 |
XM_017029342.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884831.1 |
XM_017029343.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884832.1 |
XM_017029344.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884833.1 |
XM_017029345.1 |
435 |
UTR 5 |
|
|
XP_016884834.1 |
XM_017029346.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884835.1 |
XM_017029347.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884836.1 |
XM_017029348.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884837.1 |
XM_017029349.1 |
435 |
Silent Mutation |
TAC,TAT |
Y24Y |
XP_016884838.1 |
- Gene
- INTS6L-AS1
- Gene Name
- INTS6L antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details