Product Details

SNP ID
rs150253236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129532882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTTGGAGAAAAGATATAGCATT[A/C]GCACTGAGATGGAGTGAAGAATAAG
Phenotype
MIM: 300535 MIM: 300012
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OCRL PubMed Links

Gene Details

Gene
OCRL
Gene Name
OCRL, inositol polyphosphate-5-phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000276.3 Intron NP_000267.2
NM_001318784.1 Intron NP_001305713.1
NM_001587.3 Intron NP_001578.2
XM_005262422.2 Intron XP_005262479.1
XM_011531344.2 Intron XP_011529646.1
XM_011531345.2 Intron XP_011529647.1
XM_017029554.1 Intron XP_016885043.1
Gene
SMARCA1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
There are no transcripts associated with this gene.

View Full Product Details