Product Details

SNP ID
rs150278179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47652099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCACCTCTAGCAACATGTGGAT[A/G]TGGGCTTTGATATTCATGGAGTCCT
Phenotype
MIM: 311040 MIM: 300234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELK1 PubMed Links

Gene Details

Gene
ELK1
Gene Name
ELK1, ETS transcription factor
There are no transcripts associated with this gene.

Gene
UXT
Gene Name
ubiquitously expressed prefoldin like chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004182.3 655 Silent Mutation CAC,CAT H134H NP_004173.1
NM_153477.2 655 Silent Mutation CAC,CAT H146H NP_705582.1
Gene
UXT-AS1
Gene Name
UXT antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details