Product Details

SNP ID
rs150424084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119406675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAGGAAGGGTTCCTTGCCCTTT[A/C]TCGAGGGGTTTCCCTCACTGTTGTA
Phenotype
MIM: 300641
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A43 PubMed Links

Gene Details

Gene
SLC25A43
Gene Name
solute carrier family 25 member 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145305.2 600 Missense Mutation TAT,TCT Y164S NP_660348.2
XM_017029340.1 600 Missense Mutation TAT,TCT Y164S XP_016884829.1
XM_017029341.1 600 Missense Mutation TAT,TCT Y164S XP_016884830.1

View Full Product Details